Läs mer
O-Acetyl Scopolamine Hydrobromide, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | O-Acetyl Scopolamine Hydrobromide |
| CAS | 5027-67-8 |
| Molekylformel | C19H24BrNO5 |
| Kvantitet | 50 mg |
| Synonym | 6β,7β-Epoxy-1αH,5αH-tropan-3α-ol (-)-Tropate (Ester) Acetate Hydrobromide, Scopolamine Acetate Hydrobromide, (-)-Scopolamine Acetate Hydrobromide, NSC 172233, O-Acetyl-(-)-hyoscine Hydrobromide |
| InChI formel | InChI=1S/C19H23NO5.BrH/c1-11(21)23-10-14(12-6-4-3-5-7-12)19(22)24-13-8-15-17-18(25-17)16(9-13)20(15)2;/h3-7,13-18H,8-10H2,1-2H3;1H/t13?,14-,15-,16+,17-,18+;/m1./s1 |
| LEDER | O=C([C@H](COC(C)=O)C1=CC=CC=C1)O[C@H]2C[C@H]3[C@H](O4)[C@H]4[C@@H](C2)N3C.Br |
| IUPAC-namn | [(1R,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.02,4]nonan-7-yl] (2S)-3-acetyloxy-2-phenylpropanoate;hydrobromide |
| Molekylvikt (g/mol) | 426.3 |
| Formel vikt | 425.0838 |