Läs mer
o-Acetyllamivudine, TRC
Produktkod. 30326210
Change view
Kvantitet:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30326210 250 mg
30317561 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30326210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA269810250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Acetyllamivudine
CAS 158704-07-5
Molekylformel C10 H13 N3 O4 S
Kvantitet 250 mg
Synonym o-Acetyllamivudine, 2(1H)-Pyrimidinone, 1-[(2R,5S)-2-[(acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-, 1-[(2R,5S)-2-[(Acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-2(1H)-pyrimidinone, 2(1H)-Pyrimidinone, 1-[2-[(acetyloxy)methyl]-1,3-oxathiolan-5-yl]-4-amino-, (2R-cis)-, O-Acetyllamivudine
InChI formel InChI=1S/C10H13N3O4S/c1-6(14)16-4-9-17-8(5-18-9)13-3-2-7(11)12-10(13)15/h2-3,8-9H,4-5H2,1H3,(H2,11,12,15)/t8-,9+/m0/s1
LEDER CC(=O)OC[C@@H]1O[C@@H](CS1)N2C=CC(=NC2=O)N
IUPAC-namn [(2R,5S)-5-(4-amino-2-oxopyrimidin-1-yl)-1,3-oxathiolan-2-yl]methyl acetate
Molekylvikt (g/mol) 271.293
Formel vikt 271.063