Läs mer
o-Anisylhydrazine, TRC
Produktkod. 30331518
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30331518 250 mg
1 options
Produktkod. 30331518 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA674005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material o-Anisylhydrazine
CAS 18312-46-4
Molekylformel C7 H10 N2 O
Kvantitet 250 mg
Synonym (2-Methoxyphenyl)hydrazine, (o-Methoxyphenyl)hydrazine, 2-Methoxyphenylhydrazine
InChI formel InChI=1S/C7H10N2O/c1-10-7-5-3-2-4-6(7)9-8/h2-5,9H,8H2,1H3
LEDER COc1ccccc1NN
IUPAC-namn (2-methoxyphenyl)hydrazine
Molekylvikt (g/mol) 138.167
Formel vikt 138.079