missing translation for 'onlineSavingsMsg'
Learn More

O-Benzyl-L-serine, 99%, ACROS Organics™

1097.00 SEK - 3667.22 SEK

Chemical Identifiers

CAS 4726-96-9
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
InChI Key IDGQXGPQOGUGIX-VIFPVBQESA-N
Synonym o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
PubChem CID 78457
IUPAC Name (2S)-2-amino-3-phenylmethoxypropanoic acid
SMILES C1=CC=C(C=C1)COCC(C(=O)O)N
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11995511
View Documents
Acros Organics
440930050
5g Glass bottle
1097.00 SEK
5g
View alternative products
Save Up to 
»
Estimated Shipment: 01-10-2021
Please to purchase this item. Need a web account? Register with us today!
11925521
View Documents
Acros Organics
440930250
25g Glass bottle
3667.22 SEK
25g
View alternative products
Save Up to 
»
Estimated Shipment: 01-10-2021
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

4726-96-9
195.22
o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
(2S)-2-amino-3-phenylmethoxypropanoic acid
C10H13NO3
IDGQXGPQOGUGIX-VIFPVBQESA-N
78457
C1=CC=C(C=C1)COCC(C(=O)O)N

Specifications

O-Benzyl-L-serine
Glass bottle
1% Max.
210°C to 213°C
4726-96-9
100.0
C10H13NO3
IDGQXGPQOGUGIX-VIFPVBQESA-N
(2S)-2-amino-3-phenylmethoxypropanoic acid
78457
Powder
Authentic
22.5
White
5g
98.5
98.5% min. (HPLC)
o-benzyl-l-serine, h-ser bzl-oh, s-2-amino-3-benzyloxy propanoic acid, l-serine, o-phenylmethyl, 2s-2-amino-3-benzyloxy propanoic acid, o-phenylmethyl-l-serine, serine, o-phenylmethyl, benzylserine, o-benzylserine #, z-o-benzyl-l-serine
C1=CC=C(C=C1)COCC(C(=O)O)N
195.22
195.22
99%
Safety and Handling

Safety and Handling

Product Identifier
  • O-Benzyl-L-serine

SDS
Product Certifications