Läs mer
O-Benzyl-L-tyrosine Benzyl Ester Hydrochloride, TRC
Produktkod. 30347139
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30347139 10 g
1 options
Produktkod. 30347139 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB29620510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Benzyl-L-tyrosine Benzyl Ester Hydrochloride
CAS 52142-01-5
Molekylformel C23 H23 N O3 . H Cl
Kvantitet 10 g
Synonym O-(Phenylmethyl)-L-tyrosine Phenylmethyl Ester Hydrochloride, 3-[p-(Benzyloxy)phenyl]-alanine Benzyl Ester Hydrochloride
InChI formel InChI=1S/C23H23NO3.ClH/c24-22(23(25)27-17-20-9-5-2-6-10-20)15-18-11-13-21(14-12-18)26-16-19-7-3-1-4-8-19;/h1-14,22H,15-17,24H2;1H/t22-;/m0./s1
LEDER Cl.N[C@@H](Cc1ccc(OCc2ccccc2)cc1)C(=O)OCc3ccccc3
IUPAC-namn benzyl (2S)-2-amino-3-(4-phenylmethoxyphenyl)propanoate;hydrochloride
Molekylvikt (g/mol) 397.895
Formel vikt 397.144