Läs mer
O-Benzyl-N-[(benzyloxy)carbonyl]-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-L-tyrosine Benzyl Ester, TRC
Produktkod. 30347795
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30347795 50 mg
30341462 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30347795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB23150050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Benzyl-N-[(benzyloxy)carbonyl]-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-L-tyrosine Benzyl Ester
CAS 600737-82-4
Molekylformel C37H40BNO7
Kvantitet 50 mg
Synonym O-Phenylmethyl-N-[(phenylmethoxy)carbonyl]-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-L-tyrosine Phenylmethyl Ester
InChI formel InChI=1S/C37H40BNO7/c1-36(2)37(3,4)46-38(45-36)31-22-30(20-21-33(31)42-24-27-14-8-5-9-15-27)23-32(34(40)43-25-28-16-10-6-11-17-28)39-35(41)44-26-29-18-12-7-13-19-29/h5-22,32H,23-26H2,1-4H3,(H,39,41)
LEDER O=C(OCC1=CC=CC=C1)N[C@H](C(OCC2=CC=CC=C2)=O)CC3=CC(B4OC(C)(C)C(C)(C)O4)=C(OCC5=CC=CC=C5)C=C3
IUPAC-namn benzyl (S)-3-(4-(benzyloxy)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)-2-(((benzyloxy)carbonyl)amino)propanoate
Molekylvikt (g/mol) 621.53
Formel vikt 621.29