Läs mer
o-Cresol Beta-D-Glucuronide, TRC
Produktkod. 30133564
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133564 50 mg
30005072 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC78200050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 111897-99-5
Molekylformel C13 H16 O7
Molekylvikt (g/mol) 284.26
Synonym O-Cresol-B-D-Glucuronide
IUPAC-namn (2S,3S,4S,5R,6R)-3,4,5-trihydroxy-6-(2-methylphenoxy)oxane-2-carboxylic acid
LEDER Cc1ccccc1O[C@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O
Specifikationer
Kemiskt namn eller material o-Cresol Beta-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C13H16O7/c1-6-4-2-3-5-7(6)19-13-10(16)8(14)9(15)11(20-13)12(17)18/h2-5,8-11,13-16H,1H3,(H,17,18)/t8-,9-,10+,11-,13-/m0/s1
Formel vikt 284.0896
Procent renhet >95%