Läs mer
o-Cymene, TRC
Produktkod. 30362809
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30362809 100 mg
1 options
Produktkod. 30362809 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC994100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-Methyl-2-isopropylbenzene
CAS 527-84-4
Molekylformel C10 H14
Kvantitet 100 mg
Synonym Benzene, 1-methyl-2-(1-methylethyl)-, o-Cymene (8CI), 1-Methyl-2-(1-methylethyl)benzene, 1-Isopropyl-2-methylbenzene, 1-Methyl-2-cumene, 1-Methyl-2-isopropylbenzene, 2-Isopropyltoluene, NSC 73976, o-Cymol, o-Isopropyltoluene, α-Cymene
InChI formel InChI=1S/C10H14/c1-8(2)10-7-5-4-6-9(10)3/h4-8H,1-3H3
LEDER CC(C)c1ccccc1C
IUPAC-namn 1-methyl-2-propan-2-ylbenzene
Molekylvikt (g/mol) 134.218
Formel vikt 134.11