Läs mer
O-Desethyl Azilsartan, TRC
Produktkod. 30132505
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132505 50 mg
30012879 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132505 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28925550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1442400-68-1
Molekylformel C23 H16 N4 O5
Molekylvikt (g/mol) 428.4
Synonym 2-Oxo-3-((2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-2,3-dihydro-1H-benzo[d]imidazole-4-carboxylic Acid
IUPAC-namn 2-oxo-3-[[4-[2-(5-oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]-1H-benzimidazole-4-carboxylic acid
LEDER OC(=O)c1cccc2NC(=O)N(Cc3ccc(cc3)c4ccccc4C5=NOC(=O)N5)c12
Specifikationer
Kemiskt namn eller material O-Desethyl Azilsartan
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C23H16N4O5/c28-21(29)17-6-3-7-18-19(17)27(22(30)24-18)12-13-8-10-14(11-9-13)15-4-1-2-5-16(15)20-25-23(31)32-26-20/h1-11H,12H2,(H,24,30)(H,28,29)(H,25,26,31)
Formel vikt 428.1121
Procent renhet >95%