Läs mer
O-Desethyl Candesartan, TRC
Produktkod. 30137930
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137930 25 mg -
30008105 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137930 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28204025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 168434-02-4
Molekylformel C22H16N6O3
Molekylvikt (g/mol) 412.4
Synonym 2,3-Dihydro-2-oxo-3-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-4-carboxylic Acid, 2,3-Dihydro-2-oxo-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-
LEDER OC(=O)c1cccc2NC(=O)N(Cc3ccc(cc3)c4ccccc4c5nnn[nH]5)c12
Specifikationer
Kemiskt namn eller material O-Desethyl Candesartan
Kvantitet 25 mg
InChI formel InChI=1S/C22H16N6O3/c29-21(30)17-6-3-7-18-19(17)28(22(31)23-18)12-13-8-10-14(11-9-13)15-4-1-2-5-16(15)20-24-26-27-25-20/h1-11H,12H2,(H,23,31)(H,29,30)(H,24,25,26,27)