Läs mer
O-Desethyl-O-propyl Methisosildenafil, TRC
Produktkod. 30355061
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355061 10 mg
30365880 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30355061 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD28949510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1391053-82-9
Molekylformel C24 H34 N6 O4 S
Molekylvikt (g/mol) 502.6296
Synonym rel-5-[5-[[(3R,5S)-3,5-Dimethyl-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, Propoxyphenyl Aildenafil
IUPAC-namn 5-[5-[(3R,5S)-3,5-dimethylpiperazin-1-yl]sulfonyl-2-propoxyphenyl]-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
LEDER CCCOc1ccc(cc1C2=NC(=O)c3c(N2)c(CCC)nn3C)S(=O)(=O)N4C[C@@H](C)N[C@@H](C)C4
Specifikationer
Kemiskt namn eller material Propoxyphenyl Aildenafil
Kvantitet 10 mg
InChI formel InChI=1S/C24H34N6O4S/c1-6-8-19-21-22(29(5)28-19)24(31)27-23(26-21)18-12-17(9-10-20(18)34-11-7-2)35(32,33)30-13-15(3)25-16(4)14-30/h9-10,12,15-16,25H,6-8,11,13-14H2,1-5H3,(H,26,27,31)/t15-,16+
Formel vikt 502.2362