Läs mer
O-Desisopropyl-O-methyl Bisoprolol Hemifumarate, TRC
Produktkod. 30352695
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30352695 100 mg
1 options
Produktkod. 30352695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD290190100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol
CAS 1346604-00-9
Molekylformel C16 H27 N O4
Kvantitet 100 mg
Synonym (2RS)-1-(Isopropylamino)-3-[4-(2-methoxyethoxy)methylphenoxy]propan-2-ol, Bisoprolol Fumarate Imp. Q (EP), 1-[4-[(2-Methoxyethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]-2-propanol (ACI), 2-Propanol, 1-[4-[(2-methoxyethoxy)methyl]phenoxy]-3-[(1-methylethyl)amino]- (ACI), 1-[4-(2-Methoxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
InChI formel InChI=1S/C16H27NO4/c1-13(2)17-10-15(18)12-21-16-6-4-14(5-7-16)11-20-9-8-19-3/h4-7,13,15,17-18H,8-12H2,1-3H3
LEDER COCCOCc1ccc(OCC(O)CNC(C)C)cc1
IUPAC-namn 1-[4-(2-methoxyethoxymethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol
Molekylvikt (g/mol) 297.39
Formel vikt 297.194