Learn More
O-Desmethyl Adapalene, TRC
Product Code. 30360487
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30360487 10 mg 10mg
1 options
Product Code. 30360487 Supplier Toronto Research Chemicals Supplier No. TRCD29068010MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material O-Desmethyl Adapalene
CAS 125316-60-1
Molecular Formula C27 H26 O3
Quantity 10 mg
Synonym 6-(4-Hydroxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-2-naphthalenecarboxylic Acid, 6-[3-(1-Adamantyl)-4-hydroxyphenyl]-2-naphthalenecarboxylic Acid, AHPN, CD 437, CD 437/AHPN
InChI Formula InChI=1S/C27H26O3/c28-25-6-5-22(20-1-2-21-11-23(26(29)30)4-3-19(21)10-20)12-24(25)27-13-16-7-17(14-27)9-18(8-16)15-27/h1-6,10-12,16-18,28H,7-9,13-15H2,(H,29,30)/t16-,17+,18-,27-
SMILES OC(=O)c1ccc2cc(ccc2c1)c3ccc(O)c(c3)[C@@]45C[C@@H]6C[C@@H](C[C@@H](C6)C4)C5
IUPAC Name 6-[3-(1-adamantyl)-4-hydroxyphenyl]naphthalene-2-carboxylic acid
Molecular Weight (g/mol) 398.49
Formula Weight 398.1882