Läs mer
O-Desmethyl Astemizole, TRC
Produktkod. 30011867
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011867 10 mg
30012638 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29075010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73736-50-2
Molekylformel C27 H29 F N4 O . Cl H
Molekylvikt (g/mol) 481
Synonym Desmethyl Astemizole Hydrochloride
IUPAC-namn 4-[2-[4-[[1-[(4-fluorophenyl)methyl]benzimidazol-2-yl]amino]piperidin-1-yl]ethyl]phenol;hydrochloride
LEDER Cl.Oc1ccc(CCN2CCC(CC2)Nc3nc4ccccc4n3Cc5ccc(F)cc5)cc1
Specifikationer
Kemiskt namn eller material O-Desmethyl Astemizole
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C27H29FN4O.ClH/c28-22-9-5-21(6-10-22)19-32-26-4-2-1-3-25(26)30-27(32)29-23-14-17-31(18-15-23)16-13-20-7-11-24(33)12-8-20;/h1-12,23,33H,13-19H2,(H,29,30);1H
Formel vikt 480.2092
Procent renhet >95%