Läs mer
O-Desmethyl Diltiazem Hydrochloride, TRC
Produktkod. 30135257
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135257 25 mg
30133669 50 mg
30010287 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135257 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29160525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142926-07-6
Molekylformel C21H25ClN2O4S
Molekylvikt (g/mol) 436.95
Synonym (2S,3S)-3-(Acetyloxy)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-hydroxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S-cis)-3-(Acetyloxy)-5-[2-(dimethylamino)
IUPAC-namn (2S,3S)-5-(2-(dimethylamino)ethyl)-2-(4-hydroxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
LEDER CC(O[C@@H]1[C@H](C2=CC=C(C=C2)O)SC3=CC=CC=C3N(C1=O)CCN(C)C)=O.Cl
Specifikationer
Kemiskt namn eller material rac cis-O-Desmethyl Diltiazem
Kvantitet 25 mg
InChI formel InChI=1S/C21H24N2O4S.ClH/c1-14(24)27-19-20(15-8-10-16(25)11-9-15)28-18-7-5-4-6-17(18)23(21(19)26)13-12-22(2)3;/h4-11,19-20,25H,12-13H2,1-3H3;1H/t19-,20+;/m1./s1
Formel vikt 436.12