Läs mer
O-Desmethyl Everolimus (Technical Grade), TRC
Produktkod. 30353704
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30353704 10 mg
1 options
Produktkod. 30353704 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29473010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Desmethyl Everolimus
CAS 745779-75-3
Molekylformel C52H81NO14
Kvantitet 10 mg
Synonym Desmethyl Everolimus
InChI formel InChI=1S/C52H81NO14/c1-31-15-11-10-12-16-32(2)41(55)29-39-20-18-37(7)52(62,67-39)49(59)50(60)53-22-14-13-17-40(53)51(61)66-44(34(4)27-38-19-21-43(65-24-23-54)45(28-38)63-8)30-42(56)33(3)26-36(6)47(58)48(64-9)46(57)35(5)25-31/h10-12,15-16,26,31,33-35,37-41,43-45,47-48,54-55,58,62H,13-14,17-25,27-30H2,1-9H3/b12-10+,15-11+,32-16+,36-26+/t31-,33?,34-,35-,37-,38+,39+,40+,41+,43-,44+,45-,47-,48+,52-/m1/s1
LEDER CCC#CCC#CCC#CCC#CCC#CCCCCC(=O)ON1C(=O)CCC1=O
Molekylvikt (g/mol) 944.2
Formel vikt 943.5657