Läs mer
O-Desmethyl Galanthamine, TRC
Produktkod. 30359997
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359997 10 mg
30359372 5 mg
30354146 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359997 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29203010MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60755-80-8
Molekylformel C16 H19 N O3
Molekylvikt (g/mol) 273.33
Synonym 6H-Benzofuro[3a,3,2-ef][2]benzazepine-3,6-diol, 4a,5,9,10,11,12-hexahydro-11-methyl-, (4aS,6R,8aS)-, O-Desmethyl Galanthamine, O-Desmethyl Galantamine
LEDER CN1CC[C@@]23C=C[C@H](O)C[C@@H]2Oc4c(O)ccc(C1)c34
Specifikationer
Kemiskt namn eller material O-Desmethyl Galanthamine
Kvantitet 10 mg
InChI formel InChI=1S/C16H19NO3/c1-17-7-6-16-5-4-11(18)8-13(16)20-15-12(19)3-2-10(9-17)14(15)16/h2-5,11,13,18-19H,6-9H2,1H3/t11-,13-,16-/m0/s1
Formel vikt 273.1365