Läs mer
O-Desmethyl Gatifloxacin, TRC
Produktkod. 30353533
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30353533 5 mg
1 options
Produktkod. 30353533 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2916505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Desmethyl Gatifloxacin
CAS 616205-76-6
Molekylformel C18H20FN3O4
Kvantitet 5 mg
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-(3-methyl-1-piperazinyl)-4-oxo-, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, O-Desmethylgatifloxacin, 8-Hydroxygatifloxacin
InChI formel InChI=1S/C18H20FN3O4/c1-9-7-21(5-4-20-9)15-13(19)6-11-14(17(15)24)22(10-2-3-10)8-12(16(11)23)18(25)26/h6,8-10,20,24H,2-5,7H2,1H3,(H,25,26)
LEDER CC1CN(CCN1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2O)C(=O)O
Molekylvikt (g/mol) 361.37
Formel vikt 361.1438