Läs mer
O-Desmethyl Gatifloxacin, TRC
Produktkod. 30359995
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359995 10 mg
30359852 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30359995 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29165010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Desmethyl Gatifloxacin
CAS 616205-76-6
Molekylformel C18H20FN3O4
Kvantitet 10 mg
Synonym 3-Quinolinecarboxylic acid, 1-cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-(3-methyl-1-piperazinyl)-4-oxo-, 1-Cyclopropyl-6-fluoro-1,4-dihydro-8-hydroxy-7-(3-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid, O-Desmethylgatifloxacin, 8-Hydroxygatifloxacin
InChI formel InChI=1S/C18H20FN3O4/c1-9-7-21(5-4-20-9)15-13(19)6-11-14(17(15)24)22(10-2-3-10)8-12(16(11)23)18(25)26/h6,8-10,20,24H,2-5,7H2,1H3,(H,25,26)
LEDER CC1CN(CCN1)c2c(F)cc3C(=O)C(=CN(C4CC4)c3c2O)C(=O)O
Molekylvikt (g/mol) 361.37
Formel vikt 361.1438