Läs mer
O-Desmethyl Metoclopramide Hydrochloride, TRC
Produktkod. 30005555
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005555 25 mg
30003846 10 mg
30003555 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30005555 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29209525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 38059-78-8
Molekylformel C13 H20 Cl N3 O2 . Cl H
Molekylvikt (g/mol) 322.23
Synonym Metoclopramide Ep Impurity F
IUPAC-namn 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-hydroxybenzamide;hydrochloride
LEDER Cl.CCN(CC)CCNC(=O)c1cc(Cl)c(N)cc1O
Specifikationer
Kemiskt namn eller material O-Desmethyl Metoclopramide, Hydrochloride
Kvantitet 25 mg
InChI formel InChI=1S/C13H20ClN3O2.ClH/c1-3-17(4-2)6-5-16-13(19)9-7-10(14)11(15)8-12(9)18;/h7-8,18H,3-6,15H2,1-2H3,(H,16,19);1H
Formel vikt 321.1011