Läs mer
O-Desmethyl Midostaurin, TRC
Produktkod. 17942215
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17942215 10 mg
17973055 5 mg
17923155 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17942215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29140010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 740816-86-8
Molekylformel C34 H28 N4 O4
Molekylvikt (g/mol) 556.61
Synonym N-[(9S,10R,11R,13R)-2,3,10,11,12,13-Hexahydro-10-hydroxy-9-methyl-1-oxo-9,13-epoxy-1H,9H-diindolo[1,2,3-gh:3',2',1'-lm]pyrrolo[3,4-j][1,7]benzodiazonin-11-yl]-N-methylbenzamide, CGP 62221, O-Desmethyl-PKC 412
LEDER CN([C@@H]1CC2O[C@@](C)([C@@H]1O)n3c4ccccc4c5c6CNC(=O)c6c7c8ccccc8n2c7c35)C(=O)c9ccccc9
Specifikationer
Kemiskt namn eller material O-Desmethyl Midostaurin
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C34H28N4O4/c1-34-31(39)24(36(2)33(41)18-10-4-3-5-11-18)16-25(42-34)37-22-14-8-6-12-19(22)27-28-21(17-35-32(28)40)26-20-13-7-9-15-23(20)38(34)30(26)29(27)37/h3-15,24-25,31,39H,16-17H2,1-2H3,(H,35,40)/t24-,25?,31-,34+/m1/s1
Formel vikt 556.2111
Procent renhet >95%