Läs mer
O-Desmethyl Mycophenolic Acid, TRC
Produktkod. 30362329
Change view
Kvantitet:
1 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362329 25 mg
30360989 1 mg
30358560 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362329 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29207825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31858-65-8
Molekylformel C16 H18 O6
Molekylvikt (g/mol) 306.31
IUPAC-namn (E)-6-(4,6-dihydroxy-7-methyl-3-oxo-1H-2-benzofuran-5-yl)-4-methylhex-4-enoic acid
LEDER C\C(=C/Cc1c(O)c(C)c2COC(=O)c2c1O)\CCC(=O)O
Specifikationer
Kemiskt namn eller material O-Desmethyl Mycophenolic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C16H18O6/c1-8(4-6-12(17)18)3-5-10-14(19)9(2)11-7-22-16(21)13(11)15(10)20/h3,19-20H,4-7H2,1-2H3,(H,17,18)/b8-3+
Formel vikt 306.1103