Läs mer
O-Desmethyl Pyrilamine, TRC
Produktkod. 30137215
Change view
Kvantitet:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137215 10 mg
30136715 100 mg
30006109 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29219510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57830-29-2
Molekylformel C16H21N3O
Molekylvikt (g/mol) 271.36
Synonym O-Desmethyl Mepyramine
LEDER CN(C)CCN(Cc1ccc(O)cc1)c2ccccn2
Specifikationer
Kemiskt namn eller material O-Desmethyl pyrilamine
Kvantitet 10 mg
InChI formel InChI=1S/C16H21N3O/c1-18(2)11-12-19(16-5-3-4-10-17-16)13-14-6-8-15(20)9-7-14/h3-10,20H,11-13H2,1-2H3
Formel vikt 271.1685