Läs mer
O-Desmethyl Quinidine, TRC
Produktkod. 30365926
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30365926 2.5 mg 2.5mg
30359771 10 mg -
30359552 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2921402.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70877-75-7
Molekylformel C19 H22 N2 O2
Molekylvikt (g/mol) 310.39
IUPAC-namn 4-[(S)-[(2R,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-hydroxymethyl]quinolin-6-ol
LEDER O[C@H]([C@H]1C[C@@H]2CCN1C[C@@H]2C=C)c3ccnc4ccc(O)cc34
Specifikationer
Kemiskt namn eller material O-Desmethyl Quinidine
Kvantitet 2.5 mg
InChI formel InChI=1S/C19H22N2O2/c1-2-12-11-21-8-6-13(12)9-18(21)19(23)15-5-7-20-17-4-3-14(22)10-16(15)17/h2-5,7,10,12-13,18-19,22-23H,1,6,8-9,11H2/t12-,13-,18+,19-/m0/s1
Formel vikt 310.1681