Läs mer
O-Desmethyl Tramadol Beta-D-Glucuronide (Mixture of Diastereomers), TRC
Produktkod. 30009603
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30009603 1 mg
1 options
Produktkod. 30009603 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2947351MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 383891-39-2
Molekylformel C21 H31 N O8
Molekylvikt (g/mol) 425.47
Synonym O-Desmethyltramadol Glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-6-[3-[(1S,2S)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
LEDER CN(C)C[C@@H]1CCCC[C@@]1(O)c2cccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)c2
Specifikationer
Kemiskt namn eller material O-Desmethyl Tramadol Beta-D-Glucuronide (Mixture of Diastereomers)
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C21H31NO8/c1-22(2)11-13-6-3-4-9-21(13,28)12-7-5-8-14(10-12)29-20-17(25)15(23)16(24)18(30-20)19(26)27/h5,7-8,10,13,15-18,20,23-25,28H,3-4,6,9,11H2,1-2H3,(H,26,27)/t13-,15-,16-,17+,18-,20+,21+/m0/s1
Formel vikt 425.205
Procent renhet >95%