Läs mer
O-Desmethyl Tramadol Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | O-Desmethyltramadol Hydrochloride |
| CAS | 185453-02-5 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C15 H23 N O2 . Cl H |
| Kvantitet | 10 mg |
| Synonym | (1RS,2RS)-2-[(Dimethylamino)methyl]-1-(3-hydroxyphenyl)cyclohexanol Hydrochloride, Tramadol Hydrochloride Imp. D (EP) as Hydrochloride, O-Desmethyltramadol Hydrochloride, Phenol, 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride (1:1), rel-, Phenol, 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride, rel- (9CI), Phenol, 3-[2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]-, hydrochloride, cis-, O-Demethyltramadol hydrochloride |
| InChI formel | InChI=1S/C15H23NO2.ClH/c1-16(2)11-13-6-3-4-9-15(13,18)12-7-5-8-14(17)10-12;/h5,7-8,10,13,17-18H,3-4,6,9,11H2,1-2H3;1H/t13-,15+;/m1./s1 |
| LEDER | Cl.CN(C)C[C@H]1CCCC[C@]1(O)c2cccc(O)c2 |
| IUPAC-namn | 3-[(1R,2R)-2-[(dimethylamino)methyl]-1-hydroxycyclohexyl]phenol;hydrochloride |
| Molekylvikt (g/mol) | 285.81 |
| Visa mer |