missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
O-GlcNAcase/OGA/MGEA5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32233
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
O-GlcNAcase/OGA/MGEA5 Polyclonal specifically detects O-GlcNAcase/OGA/MGEA5 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Specifikationer
| O-GlcNAcase/OGA/MGEA5 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| bifunctional protein NCOAT, FLJ11229, HEXC, hyaluronidase in meningioma, KIAA0679, MEA5FLJ23355, meningioma expressed antigen 5 (hyaluronidase), Meningioma-expressed antigen 5, NCOAT, Nuclear cytoplasmic O-GlcNAcase and acetyltransferase, OGA, O-GlcNAcase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10724 | |
| Store at 4C. Do not freeze. |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
| 1.0 mg/mL | |
| Immunohistochemistry 1:500 - 1:2000, Immunoprecipitation 2 - 10 ug/mg lysate, Immunohistochemistry-Paraffin 1:500 - 1:2000, Western Blot (Negative) | |
| NP_036347.1 | |
| OGA | |
| The immunogen this antibody was made to, maps to a region between residue 500 to 550 of human Meningioma Expressed Antigen 5 using the numbering given in entry NP_036347.1 (GeneID 10724). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering