Läs mer
O-[(Guanin-9-yl)methyl] Acyclovir, TRC
Produktkod. 30005977
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005977 100 mg
17920515 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005977 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG836410100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 166762-90-9
Molekylformel C14 H16 N10 O4
Molekylvikt (g/mol) 388.34
Synonym 6H-Purin-6-one, 9,9'-[1,2-ethanediylbis(oxymethylene)]bis[2-amino-1,9-dihydro-, 9,9'-[1,2-Ethanediylbis(oxymethylene)]bis[2-amino-1,9-dihydro-6H-purin-6-one], 9,9'-[Ethylenebis(oxymethylene)]bis(2-amino-1,9-dihydro-6H-purin-6-one), O-[(Guanin-9-yl)methyl] acyclovir
IUPAC-namn 2-amino-9-[2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethoxymethyl]-1H-purin-6-one
LEDER NC1=Nc2c(ncn2COCCOCn3cnc4C(=O)NC(=Nc34)N)C(=O)N1
Specifikationer
Kemiskt namn eller material O-[(Guanin-9-yl)methyl] Acyclovir
Kvantitet 100 mg
InChI formel InChI=1S/C14H16N10O4/c15-13-19-9-7(11(25)21-13)17-3-23(9)5-27-1-2-28-6-24-4-18-8-10(24)20-14(16)22-12(8)26/h3-4H,1-2,5-6H2,(H3,15,19,21,25)(H3,16,20,22,26)
Formel vikt 388.1356