Läs mer
O-Methyl Chlorthalidone, TRC
Produktkod. 30466363
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30466363 500 mg
30472292 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30466363 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM265165500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 96512-76-4
Molekylformel C15 H13 Cl N2 O4 S
Molekylvikt (g/mol) 352.793
Synonym 2-Chloro-5-(2,3-dihydro-1-methoxy-3-oxo-1H-isoindol-1-yl)benzenesulfonamide, 2-Chloro-5-(1-methoxy-3-oxo-1-isoindolinyl)benzenesulfonamide
IUPAC-namn 2-chloro-5-(1-methoxy-3-oxo-2H-isoindol-1-yl)benzenesulfonamide
LEDER COC1(NC(=O)c2ccccc12)c3ccc(Cl)c(c3)S(=O)(=O)N
Specifikationer
Kemiskt namn eller material O-Methyl Chlorthalidone
Kvantitet 500 mg
InChI formel InChI=1S/C15H13ClN2O4S/c1-22-15(11-5-3-2-4-10(11)14(19)18-15)9-6-7-12(16)13(8-9)23(17,20)21/h2-8H,1H3,(H,18,19)(H2,17,20,21)
Formel vikt 352.028