Läs mer
O-Methyliso Urea Tosylate Salt, TRC
Produktkod. 30484140
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30484140 5 g
1 options
Produktkod. 30484140 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3149805G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Methyliso Urea Tosylate Salt
CAS 7356-58-3
Molekylformel C9H14N2O4S
Kvantitet 5 g
Synonym O-Methylisourea p-Toluenesulfonate, Carbamimidic Acid Methyl Ester','4-Methylbenzenesulfonate, NSC 64951
InChI formel InChI=1S/C7H8O3S.C2H6N2O/c1-6-2-4-7(5-3-6)11(8,9)10;1-5-2(3)4/h2-5H,1H3,(H,8,9,10);1H3,(H3,3,4)
LEDER COC(=N)N.Cc1ccc(cc1)S(=O)(=O)O
IUPAC-namn 4-methylbenzenesulfonic acid;methyl carbamimidate
Molekylvikt (g/mol) 246.28
Formel vikt 246.0674