Läs mer
o-Nicotine, TRC
Produktkod. 30469218
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469218 10 mg
30468621 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30469218 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN41250010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23950-04-1
Molekylformel C10 H14 N2
Molekylvikt (g/mol) 162.23
Synonym Pyridine, 2-(1-methyl-2-pyrrolidinyl)-, α-Nicotine (7CI), 2-(1-Methyl-2-pyrrolidinyl)pyridine
IUPAC-namn 2-(1-methylpyrrolidin-2-yl)pyridine
LEDER CN1CCCC1c2ccccn2
Specifikationer
Kemiskt namn eller material o-Nicotine
Kvantitet 10 mg
InChI formel InChI=1S/C10H14N2/c1-12-8-4-6-10(12)9-5-2-3-7-11-9/h2-3,5,7,10H,4,6,8H2,1H3
Formel vikt 162.1157