Learn More
O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-Beta-D-galactopyranoside, TRC
Product Code. 30482896
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
30482896 250 mg 250mg
1 options
Product Code. 30482896 Supplier Toronto Research Chemicals Supplier No. TRCN505725250MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material O-Nitrophenyl 2,3,4,6-Tetra-O-acetyl-beta-D-galactopyranoside
CAS 3053-17-6
Molecular Formula C20 H23 N O12
Quantity 250 mg
InChI Formula InChI=1S/C20H23NO12/c1-10(22)28-9-16-17(29-11(2)23)18(30-12(3)24)19(31-13(4)25)20(33-16)32-15-8-6-5-7-14(15)21(26)27/h5-8,16-20H,9H2,1-4H3/t16-,17+,18+,19-,20-/m1/s1
SMILES CC(=O)OC[C@H]1O[C@@H](Oc2ccccc2[N+](=O)[O-])[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@H]1OC(=O)C
IUPAC Name [(2R,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-(2-nitrophenoxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol) 469.4
Formula Weight 469.122