Läs mer
O,O-Diethyl Thiophosphate-13C4 Ammonium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | O,O-Diethyl Thiophosphate-13C4 Ammonium Salt |
| CAS | 1330162-89-4 |
| Molekylformel | 13C4H14NO3PS |
| Kvantitet | 25 mg |
| Synonym | Phosphorothioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium Ethyl Phosphorothioate-13C4, Phosphorothioic Acid O,O-Diethyl Ester-13C4 Ammonium Salt, Ammonium O,O-Diethyl-13C4 Phosphorothioate, Ammonium O,O-Diethyl Thiophosphate-13C4 |
| InChI formel | InChI=1S/C4H11O3PS.H3N/c1-3-6-8(5,9)7-4-2;/h3-4H2,1-2H3,(H,5,9);1H3 |
| LEDER | SP(O[13CH2][13CH3])(O[13CH2][13CH3])=O.N |
| Molekylvikt (g/mol) | 191.17 |