Läs mer
O,O-Dimethyl Dithiophosphate-13C2 Ammonium Salt, TRC
Produktkod. 30135407
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135407 25 mg
17993315 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135407 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD47274225MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329610-82-3
Molekylformel 13C2H10NO2PS2
Molekylvikt (g/mol) 177.2
Synonym Phosphorodithioic acid O,O-di(methyl-13C) ester ammonium salt, [13C]Methyl ammonium phosphorodithioate, Ammonium O,O-di(methyl-13C) dithiophosphate, O,O-Di(methyl-13C) dithiophosphoric acid ammonium salt, O,O-Di(methyl-13C) ammonium dithiophosphate, O,O-Dimethyl ammonium phosphorodithioate-13C2, O,O-Dimethyl phosphorodithioic acid-13C2 ammonium salt, O,O-Dimethylphosphorodithioic acid 13C2 ammonium salt
LEDER [NH4+].[13CH3]OP(=S)([S-])O[13CH3]
Specifikationer
Kemiskt namn eller material O,O-Dimethyl Dithiophosphate-13C2 Ammonium salt
Kvantitet 25 mg
InChI formel InChI=1S/C2H7O2PS2.H3N/c1-3-5(6,7)4-2;/h1-2H3,(H,6,7);1H3