Läs mer
O,O-Dimethyl Phosphorothionate-13C2 Ammonium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | O,O-Dimethyl Phosphorothionate-13C2 Ammonium Salt |
| CAS | 1330052-30-6 |
| Molekylformel | 13C2H10NO3PS |
| Kvantitet | 25 mg |
| Synonym | Phosphorothioic Acid O,O-Dimethyl Ester-13C2 Ammonium Salt, Ammonium Methyl Phosphorothioate-13C2, Phosphorothioic Acid O,O-Dimethyl Ester-13C2 Ammonium Salt, Ammonium O,O-Dimethyl Phosphorothioate-13C2, Ammonium O,O-Dimethyl Thiophosphate-13C2, Ammonium Dimethyl Phosphorothioate-13C2 |
| InChI formel | InChI=1S/C2H7O3PS.H3N/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);1H3/i1+1,2+1; |
| LEDER | N.[13CH3]OP(=O)(S)O[13CH3] |
| IUPAC-namn | azanium;[(113C)methoxy(sulfido)phosphoryl]oxy(113C)methane |
| Molekylvikt (g/mol) | 161.13 |
| Formel vikt | 161.0186 |