Läs mer
O,O-Dimethyl Phosphorothionate-d6 Ammonium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | O,O-Dimethyl Phosphorothionate-d6 Ammonium Salt |
| CAS | 1330162-95-2 |
| Molekylformel | C2H4D6NO3PS |
| Kvantitet | 50 mg |
| Synonym | Phosphorothioic Acid O,O-Dimethyl Ester-d6 Ammonium Salt, Ammonium Methyl Phosphorothioate-d6, Phosphorothioic Acid O,O-Dimethyl Ester-d6 Ammonium Salt, Ammonium O,O-Dimethyl Phosphorothioate-d6, Ammonium O,O-Dimethyl Thiophosphate-d6, Ammonium Dimethyl Phosphorothioate-d6 |
| InChI formel | InChI=1S/C2H7O3PS.H3N/c1-4-6(3,7)5-2;/h1-2H3,(H,3,7);1H3/i1D3,2D3; |
| LEDER | SP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=O.N |
| Molekylvikt (g/mol) | 165.18 |
| Formel vikt | 165.0496 |