Läs mer
o-phenetidine hydrochloride, TRC
Produktkod. 30428565
Change view
Kvantitet:
250 mg
500 mg
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428565 500 mg
30426056 250 mg
30424573 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428565 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP321043500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 89808-01-5
Molekylformel C8H11NO . HCl
Molekylvikt (g/mol) 173.64
Synonym 2-Ethoxyphenylamine hydrochloride, 2-Ethoxyaniline hydrochloride
IUPAC-namn 2-ethoxyaniline;hydrochloride
LEDER Cl.CCOc1ccccc1N
Specifikationer
Kemiskt namn eller material o-phenetidine hydrochloride
Kvantitet 500 mg
InChI formel InChI=1S/C8H11NO.ClH/c1-2-10-8-6-4-3-5-7(8)9;/h3-6H,2,9H2,1H3;1H
Formel vikt 173.0607