missing translation for 'onlineSavingsMsg'
Learn More
Learn More
o-Phenylenediamine, Fisher BioReagents
Click to view available options
Quantity:
50 g
250 g
Packaging:
Amber Glass
Description
- A chromogenic peroxidase substrate useful for ELISA and other enzyme immunoassays
- Commonly used as a substrate for horseradish peroxidase (HRP) in ELISAs
- When oxidized, OPD produces a color change that is detectable by spectrophotometry
- Color intensity will increase over time as OPD is oxidzed by peroxidase
Specifications
Melting Point | 100°C |
Color | Cream |
Boiling Point | 256°C |
Packaging | Amber Glass |
Linear Formula | C6H4(NH2)2 |
Quantity | 50 g |
Merck Index | 15, 7396 |
Formula Weight | 108.1 |
Physical Form | Solid |
Chemical Name or Material | o-Phenylenediamine |
Safety and Handling
Product Identifier
- o-Phenylenediamine
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H301-Toxic if swallowed.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'