Läs mer
o-tert-Butyl-Alpha-chlorotoluene, TRC
Produktkod. 30345635
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30345635 50 mg
1 options
Produktkod. 30345635 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB90316050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material o-tert-Butyl-alpha-chlorotoluene
CAS 56240-38-1
Molekylformel C11H15Cl
Kvantitet 50 mg
Synonym 1-(Chloromethyl)-2-(1,1-dimethylethyl)benzene, o-(Chloromethyl)-tert-butylbenzene
InChI formel InChI=1S/C11H15Cl/c1-11(2,3)10-7-5-4-6-9(10)8-12/h4-7H,8H2,1-3H3
LEDER CC(C)(C)c1ccccc1CCl
IUPAC-namn 1-tert-butyl-2-(chloromethyl)benzene
Molekylvikt (g/mol) 182.69
Formel vikt 182.0862