Learn More
Alfa Aesar™ O-tert-Butyl-D-tyrosine, 95%
Brand:  Alfa Aesar™ H63130.03
Additional Details : CAS Number : 186698-58-8 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469648
  • 1272.68 SEK / 1g
Estimated Shipment date
from Supplier 30-01-2018
Add to basket
Specifications
Chemical Identifiers
O-tert-Butyl-D-tyrosine
237.29
H-D-Tyr(tBu)-OH; (R)-2-Amino-3-(4-tert-butoxyphenyl)propionic acid
186698-58-8
MFCD00077112
Specifications
O-tert-Butyl-D-tyrosine
Solid
237.29
H-D-Tyr(tBu)-OH; (R)-2-Amino-3-(4-tert-butoxyphenyl)propionic acid
186698-58-8
95%
MFCD00077112
1g