missing translation for 'onlineSavingsMsg'
Learn More

o-Toluenesulfonyl chloride, 80%, Thermo Scientific Chemicals

Catalog Number p-7047361
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
133-59-5
C7H7ClO2S
190.641
MFCD00024874
HDECRAPHCDXMIJ-UHFFFAOYSA-N
o-toluenesulfonyl chloride, 2-methylbenzene-1-sulfonyl chloride, toluenesulfonyl chloride, benzenesulfonyl chloride, 2-methyl, 2-toluenesulfonyl chloride, toluene-2-sulphonyl chloride, o-tolylsulfonyl chloride, o-tosyl chloride, unii-341l82e3wg, 2-methyl-benzenesulfonyl chloride
8623
2-methylbenzenesulfonyl chloride
CC1=CC=CC=C1S(=O)(=O)Cl
This item is not returnable. View return policy
133-59-5
C7H7ClO2S
190.641
MFCD00024874
HDECRAPHCDXMIJ-UHFFFAOYSA-N
o-toluenesulfonyl chloride, 2-methylbenzene-1-sulfonyl chloride, toluenesulfonyl chloride, benzenesulfonyl chloride, 2-methyl, 2-toluenesulfonyl chloride, toluene-2-sulphonyl chloride, o-tolylsulfonyl chloride, o-tosyl chloride, unii-341l82e3wg, 2-methyl-benzenesulfonyl chloride
8623
2-methylbenzenesulfonyl chloride
CC1=CC=CC=C1S(=O)(=O)Cl

o-Toluenesulfonyl chloride is used in organic synthesis and saccharin production. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluenesulfonyl chloride is used in organic synthesis and saccharin production. It is also used as pharmaceutical intermediate.

Solubility
Soluble in hot alcohol, ether and benzene. Reacts with water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Store under inert gas. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Melting Point 10°C
Density 1.338
Boiling Point 154°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.556
Quantity 5 g
UN Number UN3265
Beilstein 973180
Sensitivity Moisture sensitive
Solubility Information Soluble in hot alcohol,ether and benzene. Reacts with water.
Formula Weight 190.65
Percent Purity 97%
Chemical Name or Material o-Toluenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'