Läs mer
o-Tolylglyoxylic Acid Methyl Ester, TRC
Produktkod. 30453376
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30453376 100 mg
1 options
Produktkod. 30453376 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT536700100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material o-Tolylglyoxylic Acid Methyl Ester
CAS 34966-54-6
Molekylformel C26 H33 N O6
Kvantitet 100 mg
Synonym (2-Methylphenyl)glyoxylic Acid Methyl Ester, Methyl 2-(2-Methylphenyl)-2-oxoacetate, Methyl 2-Methylphenylglyoxylate, Methyl o-Toluoylformate
InChI formel InChI=1S/C26H33NO6/c1-7-30-23(28)22-21(27(26(5,6)32-22)24(29)33-25(2,3)4)19-13-15-20(16-14-19)31-17-18-11-9-8-10-12-18/h8-16,21-22H,7,17H2,1-6H3/t21-,22?/m0/s1
LEDER CCOC(=O)C1OC(C)(C)N([C@H]1c2ccc(OCc3ccccc3)cc2)C(=O)OC(C)(C)C
IUPAC-namn 3-O-tert-butyl 5-O-ethyl (4S)-2,2-dimethyl-4-(4-phenylmethoxyphenyl)-1,3-oxazolidine-3,5-dicarboxylate
Molekylvikt (g/mol) 455.543
Formel vikt 455.231