Läs mer
O-Triphenylmethoxy Quetiapine, TRC
Produktkod. 30449910
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30449910 250 mg
1 options
Produktkod. 30449910 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT809040250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material O-Triphenylmethoxy Quetiapine
CAS 844639-06-1
Molekylformel C40 H39 N3 O2 S
Kvantitet 250 mg
Synonym Dibenzo[b,f][1,4]thiazepine','11-[4-[2-[2-(triphenylmethoxy)ethoxy]ethyl]-1-piperazinyl]-, 11-[4-[2-[2-(Triphenylmethoxy)ethoxy]ethyl]-1-piperazinyl]dibenzo[b,f][1,4]thiazepine, 11-[4-[2-(2-Trityloxyethoxy)ethyl]piperazin-1-yl]dibenzo[b,f][a,4]thiazepine, 11-[4-[2-[2-(Triphenylmethoxy)ethoxy]ethyl]piperazin-1-yl]dibenzo[b,f][1,4]thiazepine, Quetiapine Fumarate Imp. O (EP)
InChI formel InChI=1S/C40H39N3O2S/c1-4-14-32(15-5-1)40(33-16-6-2-7-17-33,34-18-8-3-9-19-34)45-31-30-44-29-28-42-24-26-43(27-25-42)39-35-20-10-12-22-37(35)46-38-23-13-11-21-36(38)41-39/h1-23H,24-31H2
LEDER C(COC(c1ccccc1)(c2ccccc2)c3ccccc3)OCCN4CCN(CC4)C5=Nc6ccccc6Sc7ccccc57
IUPAC-namn 6-[4-[2-(2-trityloxyethoxy)ethyl]piperazin-1-yl]benzo[b][1,4]benzothiazepine
Molekylvikt (g/mol) 625.82
Formel vikt 625.2763