missing translation for 'onlineSavingsMsg'
Learn More

o-Vanillin, 99%, Thermo Scientific Chemicals

Catalog Number p-212055
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
148-53-8
C8H8O3
152.15
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(C=O)=C1O
This item is not returnable. View return policy
148-53-8
C8H8O3
152.15
MFCD00003322
JJVNINGBHGBWJH-UHFFFAOYSA-N
3-methoxysalicylaldehyde, o-vanillin, 6-formylguaiacol, 2-vanillin, o-vanilline, benzaldehyde, 2-hydroxy-3-methoxy, orthovanilline, 2-hydroxy-m-anisaldehyde, 6-formyl-2-methoxyphenol, 3-methoxy-2-hydroxybenzaldehyde
8991
CHEBI:78339
2-hydroxy-3-methoxybenzaldehyde
COC1=CC=CC(C=O)=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 40.0°C to 42.0°C
Color Brown to Yellow
Boiling Point 265.0°C to 266.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 10 g
Beilstein 08, IV, 1747
Solubility Information Solubility in water: slightly soluble. Other solubilities: very soluble in alcohol,ether,benzene,carbon,disulfide
Formula Weight 152.15
Percent Purity 99%
Physical Form Low Melting Solid
Chemical Name or Material o-Vanillin, 99%
Show More Show Less
compliance-icons
Product Identifier
  • o-Vanillin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'