missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
O6BTG-octylglucoside, MedChemExpress
Description
O6BTG-octylglucoside is a potent O6-methylguanine-DNAmethyl-transferase (MGMT) inhibitor, with IC50s of 32 nM in vitro (cell extracts) and 10 nM in HeLa S3 cells.
Specifications
Specifications
| Kemiskt namn eller material | O6BTG-octylglucoside |
| CAS | 382607-78-5 |
| Färg | Off-White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C24H34BrN5O7S |
| Kvantitet | 10 mg |
| Synonym | Glucose-conjugated MGMT inhibitor |
| Löslighetsinformation | DMSO : 100 mg/mL (162.20 mM; Need ultrasonic) |
| LEDER | [H][C@]1([C@]([H])(O[C@@](OCCCCCCCCN2C3=C(N=C2)C(OCC4=CC(Br)=CS4)=NC(N)=N3)([C@](O)([C@]1(O)[H])[H])[H])CO)O |
| Molekylvikt (g/mol) | 616.53 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?