missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14214-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody recognizes isoform p71 and isoform p69 of OAS2.
This gene encodes a member of the 2-5A synthetase family, essential proteins involved in the innate immune response to viral infection. The encoded protein is induced by interferons and uses adenosine triphosphate in 2′-specific nucleotidyl transfer reactions to synthesize 2′,5′-oligoadenylates. These molecules activate latent RNase L, which results in viral RNA degradation and the inhibition of viral replication. The three known members of this gene family are located in a cluster on chromosome 12. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
OAS2 | |
Polyclonal | |
Unconjugated | |
OAS2 | |
(2 5)oligo(A) synthase 2, 2 5 oligoadenylate synthase 2, 2 5A synthase 2, OAS2, p69 OAS / p71 OAS, p69OAS / p71OAS | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4939 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P29728 | |
OAS2 | |
OAS2 Fusion Protein Ag5436 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |