missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
OASIS/CREB3L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31017
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
OASIS/CREB3L1 Polyclonal specifically detects OASIS/CREB3L1 in Human samples. It is validated for Western Blot.
Specifikationer
| OASIS/CREB3L1 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 10% Glycerol with 0.01% Thimerosal | |
| CREB3L1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human CREB3L1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 90993 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance | |
| Rabbit | |
| 57 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Mouse (85%) and Rat (85%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering