Läs mer
OASIS/CREB3L1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00039PCP
Produktkod. 18678145
  • 3835.00 SEK / 0.10ml
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
OASIS/CREB3L1 Polyclonal antibody specifically detects OASIS/CREB3L1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifikationer
Specifikationer
OASIS/CREB3L1
Polyclonal
PBS
Rabbit
0.1 mL
Store at 4C in the dark.
ELISA
PerCP
BBF-2 homolog, cAMP responsive element binding protein 3-like 1, cAMP-responsive element-binding protein 3-like protein 1, OASIScyclic AMP-responsive element-binding protein 3-like protein 1, old astrocyte specifically induced substance, Old astrocyte specifically-induced substance
Produced in rabbits immunized with purified, recombinant Human OASIS/CREB3L1 (Uniprot#: Q96BA8-1; Glu396- Ser519)
90993