Läs mer
Obeticholic Acid Methyl Ester, TRC
Produktkod. 30240828
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240828 500 mg
30240499 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240828 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO099130500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 951694-73-8
Molekylformel C27 H46 O4
Molekylvikt (g/mol) 434.65
Synonym (3α,5β,6α,7α)-6-ethyl-3,7-dihydroxy-cholan-24-oic acid methyl ester, Methyl 3α,7-dihydroxy-6-ethyl-5β-cholanate, Obeticholic Acid Methyl Ester
IUPAC-namn methyl (4R)-4-[(3R,5S,6R,7R,8S,9S,10S,13R,14S,17R)-6-ethyl-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate
LEDER CC[C@H]1[C@@H](O)[C@H]2[C@@H]3CC[C@H]([C@H](C)CCC(=O)OC)[C@@]3(C)CC[C@@H]2[C@@]4(C)CC[C@@H](O)C[C@@H]14
Specifikationer
Kemiskt namn eller material Obeticholic Acid Methyl Ester
Kvantitet 500 mg
InChI formel InChI=1S/C27H46O4/c1-6-18-22-15-17(28)11-13-27(22,4)21-12-14-26(3)19(16(2)7-10-23(29)31-5)8-9-20(26)24(21)25(18)30/h16-22,24-25,28,30H,6-15H2,1-5H3/t16-,17-,18-,19-,20+,21+,22+,24+,25-,26-,27-/m1/s1
Formel vikt 434.3396