Läs mer
Obidoxime Chloride, TRC
Produktkod. 30413118
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30413118 1 g
1 options
Produktkod. 30413118 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO1000001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Obidoxime Chloride
CAS 114-90-9
Molekylformel C14 H16 N4 O3 . 2 Cl
Kvantitet 1 g
Synonym Pyridinium','1,1'-[oxybis(methylene)]bis[4-[(hydroxyimino)methyl]-','chloride (1:2), 1,1'-(Oxydimethylene)bis[4-formylpyridinium chloride]','dioxime (7CI), Pyridinium','1,1'-(oxydimethylene)bis[4-formyl-','dichloride','dioxime (8CI), Pyridinium','1,1'-[oxybis(methylene)]bis[4-[(hydroxyimino)methyl]-','dichloride (9CI), 1,1'-(oxydimethylene)bis[4-formylpyridinium] dichloride dioxime, BH 6, Bis(4-hydroxyiminomethylpyridiniummethyl) ether dichloride, Efosim, LuH6, LuH6-Cl2, LueH 6, Obidoxime chloride, Obidoxime hydrochloride, Toksobidin, Toxobidin, Toxogonin, Toxogonin dichloride, Toxogonine
InChI formel InChI=1S/C14H14N4O3.2ClH/c19-15-9-13-1-5-17(6-2-13)11-21-12-18-7-3-14(4-8-18)10-16-20;;/h1-10H,11-12H2;2*1H
LEDER [Cl-].[Cl-].O\N=C\c1cc[n+](COC[n+]2ccc(\C=N\O)cc2)cc1
IUPAC-namn (NE)-N-[[1-[[4-[(E)-hydroxyiminomethyl]pyridin-1-ium-1-yl]methoxymethyl]pyridin-1-ium-4-yl]methylidene]hydroxylamine;dichloride
Molekylvikt (g/mol) 359.21
Formel vikt 358.0599